VWR® Pocket Thermometers
À propos de cet article
Pocket glass thermometers feature permanently fused markings.
- Red spirit-filled
- Includes a heavy-duty case
- Compact size
- Glass fabrication
- Permanently fused markings
Markings are easy to read against thermometer background.
Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.
4 Options Available Below
Product Details & Documents
Pocket glass thermometers feature permanently fused markings.
- Red spirit-filled
- Includes a heavy-duty case
- Compact size
- Glass fabrication
- Permanently fused markings
Markings are easy to read against thermometer background.
Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.
Documents
Product Family Options
Thermometers
Informations sur le produit
- RangeLongueurDivisionsPackagingAvailabilityPrice
- −35…+50 °C160 mm (6⁵⁄₁₆")1 °CClosed metal caseDiscontinued-
- −10...+110 °C (0...+220 °F)160 mm (6⁵⁄₁₆")1 °C, 2 °FClosed metal caseDiscontinued-
- −35…+50 °C160 mm (6⁵⁄₁₆")1 °CClosed plastic caseDiscontinued-
- −10...+110 °C (0...+220 °F)160 mm (6⁵⁄₁₆")1 °C, 2 °FClosed plastic caseDiscontinued-
- −35…+50 °C160 mm (6⁵⁄₁₆")1 °CWindow plastic caseDiscontinued-
- −10...+110 °C (0...+220 °F)160 mm (6⁵⁄₁₆")1 °C, 2 °FClosed plastic caseDiscontinued-
- −35…+50 °C160 mm (6⁵⁄₁₆”)1 °CWindow plastic caseLog in pour la disponibilitéCase of 25Case of 25 - $1,226.76$1,226.76Prix promotionnel pour les nouveaux clientsEach$49.07Case of 25$1,226.76
- −35…+50 °C160 mm (6⁵⁄₁₆")1 °CWindow plastic caseEach$94.96Case of 25$1,704.16
- −10...+110 °C160 mm (6⁵⁄₁₆”)1 °CWindow plastic caseLog in pour la disponibilitéCase of 25Case of 25 - $883.15$883.15Prix promotionnel pour les nouveaux clientsEach$35.33Case of 25$883.15
- −5…+45 °C160 mm (6⁵⁄₁₆”)0.5 °CWindow plastic caseLog in pour la disponibilitéCase of 25Case of 25 - $826.63$826.63Prix promotionnel pour les nouveaux clientsEach$33.07Case of 25$826.63
Services
Informations sur le produit
- Catalog #DescriptionIncludesAvailabilityPrice
- 10186-100Customer Specified 1 Point Re-Calibration (0°C-100°C)ISO17025 NIST certification, customer specified 1 temperature point re-calibration & no extra shipping feeLog in pour la disponibilitéEach - $186.82