VWR® Maximum/Minimum Thermometers
# de catalogue 82021-174
Fournisseur: VWR International
Spécifications
- Accuracy:1 °C/2 °F
- Dimensions:200 x 230 mm
- Description:Dial
- Cat. no.:82021-174
- Divisions:1 °C/2 °F
- Range:−30…+60 °C (−20…+140 °F)
Spécifications
A propos de cet article
Organic liquid thermometer features a U-shaped tube mounted on a plastic casing with metal floats at the top of each column.
- Easy-to-understand dial thermometer
- Minimum and maximum temperature floats are reset easily
One float remains at the maximum temperature while the other float remains at the minimum temperature
Dial thermometer displays the current temperature and registers the maximum and minimum temperatures attained since the last reset.
Reset the floats by pushing the reset button.
Customers Who Bought This Also Bought