Stainless Steel Undermount Sink, Just Manufacturing
# de catalogue 83007-406
Fournisseur: Just Manufacturing
Small Business Enterprise
Spécifications
- Depth:7½"
- Length:21"
- Width:18"
- Cat. no.:83007-406
- Supplier No.:US1821AGR
Spécifications
A propos de cet article
This undermount sink features seamless die-drawn construction of 18-gauge, Type 304 stainless steel.
- Interior surfaces are polished to a nonporous hand-blended finish
- Fully-coated underside insulates for sound and reduces condensation
- Straight-sided compartment provides greater capacity
Certifications: Conforms to ASME/ANSI A112.19.3M.
Customers Who Bought This Also Bought