BASIKBench® Storage Drawers, Kewaunee®
Catalog # KL447051
Supplier: Avantor
Product available on GSA Advantage®
Specifications
- Color:—
- Description:Mounting bracket
- Size:—
- Cat. no.:KL447051
- Supplier No.:447051
Specifications
About this item
Lockable storage drawers feature steel construction and recessed handles and can be stacked or mounted under worksurfaces.
- Drawers feature Contour™ front styling
- The box drawer is suspended with two-member ball bearing slides
- The file drawer is suspended with three-member ball bearing slides
A mounting bracket (sold separately) is required for each hanging drawer configuration that attaches directly to the worksurface. Drawers can be keyed alike, differently, by workstation, or by area.
Customers Who Bought This Also Bought