VWR® Atlas End-Mounted Shelving
Supplier: VWR International
Certificates
About this item
Single shelves for single-side applications are powder-coated with high-quality chemical-resistant epoxy powder and constructed of 16-gauge steel.
- 16-gauge steel construction
- Coated with chemical-resistant epoxy powder
- Infinitely adjustable along vertical Atlas uprights
Color: Light neutral or snow white
Ordering information: Supplied with mounting hardware.
Customers Who Bought This Also Bought