VWR® Atlas Slot-Mounted Shelving, Double-Sided
Slot-mounted shelving for double-sided applications.
- Adjustable with use of uprights
Used with slotted uprights (see VWRSU24 series) for shelf adjustment in 2.5 cm (1") increments.
Ordering information: Two shelves are required; each is sold separately. Brackets are included.
Customers Who Bought This Also Bought