VWR® Atlas Slot-Mounted Shelving, Single-Sided
Supplier: VWR International
Certificates
About this item
Slot-mounted shelving for single-sided applications.
- Color: Light neutral
Slot-mounted shelving is used with slotted uprights (see VWRSU24 series) for shelf adjustment in 2.5 cm (1") increments.
Customers Who Bought This Also Bought