VWR® Atlas Center-Mounted Umbrella Shelving
Umbrella shelves are mounted at the top of uprights and replace a cross brace when used.
- 16-gauge steel construction
- Coated with chemical-resistant epoxy powder
Shelves are powder-coated with high-quality chemical-resistant epoxy powder and constructed of 16-gauge steel.
Ordering information: Shelf brackets are available separately; see VWRBHC12 series.
Customers Who Bought This Also Bought