Angle-Sided Shelves, TrippNT
Fournisseur: Avantor
Certificats
A propos de cet article
These PVC shelves feature angled sides and built-in safety lips on the front edges.
- Shelves are chemical-resistant and will not scratch glassware
- Can be easily mounted onto a wall
Color: White
Ordering information: Shelves are also available in safety yellow or red; contact your Avantor representative for more information.
Customers Who Bought This Also Bought