VWR® Atlas Metal Shelving, Single Lip
Supplier: VWR International
Certificates
About this item
Designed to be infinitely adjustable along Atlas uprights.
- Standard shelves are powder-coated with high-quality, chemical-resistant epoxy powder
- Constructed of 16-gauge steel
Lipped shelves can be used with the lip in the front or back and can be switched in the field. Light neutral shelves are 3 cm (13/16") thick with 1.1 cm (7/16") high lips.
Ordering information: Shelf brackets are available separately; see VWRBHC12 series.
Customers Who Bought This Also Bought