Wall-Mounted Shelf, Stainless Steel, Advance Tabco®
Fournisseur: Advance Tabco
Certificats
A propos de cet article
Constructed of strong, 18-gauge stainless steel.
- Limit to dry storage
- Back is turned up
- Entire unit welded for strength and durability
- Front and sides feature a rolled rim edge
- Mounting hardware not supplied
Extra deep 46 cm (18") shelf provides increased storage capacity.
Customers Who Bought This Also Bought