Spécifications
- Description:M13KO7 Helper Phage (Liquid)
- Size:100 µL
- Cat. no.:CA95017-590L
- Supplier No.:27-1524-01
Spécifications
A propos de cet article
M13 phage for the GST expression system.
- For phage rescue
- Colorless to light yellow liquid
- Concentration >5×10¹⁰ pfu/ml
The M13KO7 helper phage is an M13mp1 derivative containing a modified gene 11 (G changed to T at position 6,125, giving a met to ile change at codon 40 of the gene 11 protein), an insertion of the origin of replication of p15A and the kanamycin resistance gene from Tn903 at the Ava I site (position 5,825).