Freestanding Gowning Racks with Shelf, Eagle MHC
Supplier: Avantor
Certificates
About this item
Freestanding gowning rack incorporates wire shelving components that offer a more economical rack option for less demanding environments.
- Features hanger slots on 2¹/₂" centers, 24" wide
Open wire construction facilitates laminar flow. Convenient for storage of lab coats, uniforms and garments.
Customers Who Bought This Also Bought