VWR® Contour™ Double-Tiered Upright Support
# de catalogue CWW-0201-1A
Fournisseur: VWR International
Spécifications
- Width:12½>"
- Color:Snow white
- Description:Double-tiered upright support
- Height:24½"
- Material:Steel
- Cat. no.:CWW-0201-1A
Spécifications
A propos de cet article
The double tiered upright support has an intermediate shelf option for additional storage.
- Color: Light neutral
The shelf support holes are on 5 cm (2") centers to allow easy adjustment.
Customers Who Bought This Also Bought