VWR® Atlas Single Cross Brace
Fournisseur: VWR International
Certificats
A propos de cet article
Atlas cross braces are necessary to stabilize an Atlas shelf assembly.
- Color: Light neutral
One cross brace is required for all single- and double-sided assemblies.
Customers Who Bought This Also Bought