Compressed Breathing Air Analysis Kit, Sensidyne®
Fournisseur: Sensidyne
Certificats
A propos de cet article
Convenient, economical system tests the quality of compressed breathing air
- The flowmeter is adjustable from 50 to 600cc/minute
- System accepts up to 3000psi with a standard No. 346 CGA fitting
Each tube (available separately) is designed to detect a different contaminant. The reagents in the tube will change color and react quantitatively to provide a length-of-stain indication. The farther the color-stain travels along the tube, the higher the concentration.
Measures up to five common breathing air contaminants. System consists of a back pressure compensated flowmeter/regulator and tube holder, supplemented with direct-reading detector tubes.
Ordering information: Kit includes a regulator, flow controller, and tube holder. All tubes must be ordered separately.
Spécifications
- Catalog No:
- 13502-660
- 32011-233
- 14233-747
- 14233-753
- 14233-749
- 14233-751
- Measurement range:
- 5–100 ppm
- 20–160 mg/m3
- 2–24%v
- 0.3–1.5 mg/m3
- 100–3000 ppm