VWR® Contour™ Standing Height Corner Base Cabinet
Fournisseur: VWR International
Certificats
A propos de cet article
Heavy-gauge steel full-frame construction.
- Smooth, easy-to-maintain surfaces
- Phosphate-coated with a baked, chemical-resistant, powder-coat finish
- Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
Cabinet has one cupboard and includes sink-support assembly.
Two rear panels 317/8" each, two front panels 211/4" each
Clearance width 151/16"
Customers Who Bought This Also Bought