VWR® Talon® Adjustable Cylinder Stand
Catalogue # 97035-540
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Description:VWR® Talon® Adjustable Cylinder Stand
- Dimensions:47L x 19.1W cm (181/2 x 71/2")
- Cat. no.:97035-540
Spécifications
À propos de cet article
Heavy-duty, cast aluminum stand with adjustable L-brackets holds cylinders from 15.2 – 23.5 cm (6 – 91/4") in diameter.
- Prevents accidental tipping
- Fully adjustable stand
- Holds a range of cylinder sizes
Hinges open for installation without lifting the cylinder or disturbing connections.