VWR® Talon® Cylinder Stand
# de catalogue 97035-538
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Dimensions:46.4L x 15.2W (181/4 x 6")
- Description:VWR® Talon® Cylinder Stand
- Cat. no.:97035-538
Spécifications
A propos de cet article
Cast aluminum stand hinges open and has four thumbscrews to tightly grip cylinders from 20.3 – 22.9 cm (8 – 9") in diameter.
- Rugged design
- Cast aluminum construction
- Easily adjustable
The Talon® Cylinder Stand provides a very stable base of support for cylinders in a range of sizes.
Customers Who Bought This Also Bought