VWR® Talon® Gas Cylinder Stand
# de catalogue 60141-000
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Description:VWR® Talon® Gas Cylinder Stand
- Cat. no.:60141-000
Spécifications
A propos de cet article
Permits convenient, safe cylinder transfer and upright support.
- Heavy-duty cast iron construction
- Rubber support wheels
- Fully mobile
- Secure fit around a range of tank diameters
Designed in two halves that fit securely around any tank from 17.8 – 23.5 cm (7 – 91/4") in diameter.
Made of heavy-duty cast iron, support has rubber wheels that convert it into a dolly for complete mobility. Can be tipped to approximately 35° and returned to vertical position.
Customers Who Bought This Also Bought