Safety Shower Stations, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Designed to provide complete personal protection, these corrosion-resistant safety stations feature a single water inlet that supplies both the shower and eye wash.
- Supplied with rigid stainless steel pull rods
- Self-regulating flow controls are positioned inside each spray head
- Stations are provided with an ANSI-compliant sign
- 2 versions available: Orange ABS plastic shower head and galvanized steel pipes and fittings with orange PE covers or cast brass showerhead and white epoxy/polyester hybrid powder coated brass pipe and fittings
The high volume deluge shower head has a minimum 75.7 L/min. (20 gpm) flow and 50.8 cm (20") spray pattern.
Certifications: Meets ANSI Z358.1 standard.
Customers Who Bought This Also Bought