Eyesaver® Eye/Face Wash Station, Speakman®
Fournisseur: Speakman
Certificats
A propos de cet article
These wall-mounted eye/face washes feature dual spray heads activated by a stainless steel push handle.
The spray heads feature automatic flow controls and flip-top dust covers.
Certifications: Meet ANSI Z358.1 standard.
Customers Who Bought This Also Bought