Eyesaver® Multi-Spray Eye/Face Washes, Speakman®
Fournisseur: Speakman
Certificats
A propos de cet article
Eye/face washes provide six aerated sprays for gentle yet thorough rinsing of the eyes and face.
- Eye/face washes are available with wall or pedestal mount
- Bowls are available in stainless steel or plastic
Featuring an in-line strainer, the washes have a stay-open ball valve and a large rectangular bowl.
Certifications: Meet ANSI Z358.1 standard.
Customers Who Bought This Also Bought