Eyesaver® Eyewash Station, Speakman®
Fournisseur: Speakman
Certificats
A propos de cet article
For use in areas where workers could be exposed to harmful contaminants.
- Feature twin aerated spray heads and stay-open ball valve
Eyewashes flush liquid and dust contaminants from the eyes when activated by the stainless steel push handle.
Certifications: Meet ANSI Z358.1 standard.
Customers Who Bought This Also Bought