VWR® Contour™ Standing Height Base Cabinet Double Door Cupboard
Fournisseur: VWR International
Certificats
A propos de cet article
Heavy-gauge steel full-frame construction.
- Smooth, easy-to-maintain surfaces
- Phosphate-coated with a baked, chemical-resistant, powder-coat finish
- Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
- Light neutral color
A 2.54 cm (1") thick work surface creates an overall height of 91.4 cm (36"). Cupboard cabinets are supplied with one full-width adjustable shelf.
Customers Who Bought This Also Bought