Aluminum Column Tags, Ohio Valley
# de catalogue 21507-976
Fournisseur: Ohio Valley
Small Business Enterprise
Spécifications
- Description:Aluminum column tag
- Cat. no.:21507-976
- Supplier No.:2-1001
Spécifications
A propos de cet article
Designed to be attached to the column for permanent identification.
Tags can be engraved using a vibrator or simply with a ball point pen.
Customers Who Bought This Also Bought