Junkin™ Safety Confined Space Stretcher
# de catalogue 56612-680
Fournisseur: Junkin
Small Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Junkin* Safety Confined Space Stretcher
- Cat. no.:56612-680
- Supplier No.:JSA-300CS
Spécifications
A propos de cet article
Designed for rescues where space is tight.
- Lightweight 48.2 cm (19") wide stretcher features rigid steel construction
- Ample basket is 18 gauge, 2.5 cm (1") hexagon mesh netting
- Nylon web straps secure victim at chest, abdomen, thigh and calf
WxLxH:46.6x 207x19.6 (18⅜x81½x7¾")
10.4 kg (23 lbs.)
Customers Who Bought This Also Bought