VWR® Heated Recirculator, Model 1104
Fournisseur: VWR International
CE Compliant
Certificats
A propos de cet article
This small, low-cost recirculator handles applications such as thawing plasma and tempering photographic solutions.
- VWR® Two-Year Limited Warranty
Features stainless steel, PVC, brass, and nylon wetted parts. Low-liquid cutoff and over-temperature cutoff. The sealed reservoir can control closed loop systems only.
Ordering information: Supplied with 1.8m (6') power cord with grounded plug.
Caution: Note 13271-063 is 240 V, 50 Hz
Spécifications
- Catalog No:
- 13271-063
- 13271-060
- Heater:
- 750W
- 750W
- Pump port:
- 1.3 cm (1/2") NPT
- 1.3 cm (1/2") NPT
- Stability:
- ±0.2°C
- ±0.2°C
- Exterior dimensions:
- 24.1W x 20D x 20.3H cm (91/2 x 8 x 8")
- 24.1W x 20D x 20.3H cm (91/2 x 8 x 8")