Shorty Disposable Serological Pipettes, Glass, Sterile, Plugged, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Calibrated to deliver with blowout after free flow of liquid has ended
Ideal for use in tissue culture work and other procedures performed under a hood or in other confined areas.
Ordering information: Available individually wrapped or in multi-bag packs.
Customers Who Bought This Also Bought