Sink Crumb Cup Strainers, Just Manufacturing
Fournisseur: Just Manufacturing
Certificats
A propos de cet article
Crumb cup strainer has removable stainless steel basket with rubber seat stopper.
- Made of 316 type stainless steel
Tailpiece dimensions: Ext.Ø 3.8 cm and lenght 10.1 cm (1¹/₂×4").
Delivery: Strainer 58200-530 comes with chrome-plated brass tailpiece. For extra corrosion resistance, 58200-528 offers strainer, basket, and tailpiece in Type 316 stainless steel.
Customers Who Bought This Also Bought