Double Compartment Sink without Ledge, Just Manufacturing
# de catalogue 58200-516
Fournisseur: Just Manufacturing
Small Business Enterprise
Spécifications
- Depth:7½"
- Cat. no.:58200-516
- Exterior Dimensions:48.2×93.9 cm (19×37")
- Supplier No.:D-1937-A-GR
Spécifications
A propos de cet article
Fabricated of Type 304, 18-8 stainless steel
This self-rimming double sink with polished interior and top surface is thoroughly sound deadened. Punched-in center receives cup strainer. Constructed without faucet ledge to fit narrow counters.
Customers Who Bought This Also Bought