Balance Table, Waller Brothers
Catalog # VF7-6590-00
Supplier: Waller Brothers Stone
Specifications
- Description:Balance table
- Cat. no.:VF7-6590-00
- Weight:475 lbs.
- Supplier No.:STD.BAL.TBL
- W×D:35×24"
Specifications
About this item
This rock-steady weighing surface is ideal for delicate microbalance procedures.
- Balance table is made of heavy 7.6 cm (3") thick black KEMROCK® slabs
- Connected by a steel pipe with flanges
The weight of the table minimizes floor vibrations that would be transferred to an ordinary tabletop.
Customers Who Bought This Also Bought