SP Bel-Art Vibrasorb® Premium Vibration Damping Mounts, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Vibrasorb® Vibration Damping Mounts facilitate optimal use and help protect electromechanical and optical equipment from excessive wear and even structural damage that can be caused by exposure to vibration.
- Polished surface resists scratches and chemicals
- Terrazzo construction
- Non-skid neoprene cushion feet
- Support up to 16 kg (35 lbs.)
Molded of dense black and white terrazzo, these 6.4 cm (2½") thick platforms are supported on four Vibro-Absorber™ cushions with neoprene feet and support up to 16kg (35 lbs.).
The larger size will absorb vibrations down to 13Hz with a 16kg (35 lbs.) load, while the smaller size will absorb vibrations down to 16Hz. with a 16kg (35 lbs.) load. With lighter loads, the minimum frequency of damping for both sizes will increase to a value between 16Hz. and 25Hz. Both sizes feature a polished surface that resists scratches and chemical attack.
Customers Who Bought This Also Bought