VWR® Pipette Fillers
Fournisseur: VWR International
CE Compliant
Certificats
A propos de cet article
Pipette fillers do away with the frequently hazardous practice of drawing liquids into pipettes by mouth.
- 100% natural rubber construction
- For use with 1 to 45 ml pipettes
- Four color options available
They work safely with toxic, infectious, odoriferous, radioactive, corrosive, or sterile liquids.
Pipettes are made out of 100% natural rubber with three stainless steel ball valves. For use with pipettes of 1 to 45 ml capacity and 4 to 8 mm outside diameter. A volume of 100 ml is attainable with repeated evacuation. With a simple three-step process, one can eliminate dangerous mouth pipetting.
Also useful for skimming off liquids floating above a chemical precipitate or sediment.
Customers Who Bought This Also Bought