Samco™ Disposable Transfer Pipettes, Fine Tip, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These fine-tip polyethylene transfer pipettes are excellent for the precise dispensing of small drops (45 to 50 per ml) into microbiological Int.Ø.
Strips, ELISA plates, and microcentrifuge and precipitation tubes. Ideal for removing cell layers, filling electrophoresis wells, spotting chromatography plates, and other similar applications.
Specifications
- Catalog No:
- 14670-329
- 73990-068
- 14670-330
- 14670-325
- 14670-327
- 73990-052
- 14670-331
- 73990-072
- 14670-332
- 73990-070
- 14670-333
- 73990-078
- 14670-336
- 14670-339
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)