Heavy-Duty Storage Cabinets, SECURALL®
Fournisseur: Securall Cabinets
Certificats
A propos de cet article
Constructed of 18-gauge galvanized steel, these heavy-duty storage cabinets feature two double-wall doors and a locking, flush-mount handle to prevent unauthorized entry.
- Custom interiors are available to fit the specific needs
- 18-gauge all-welded extra strength construction throughout
Cabinets have the SUR-LOC™ adjustable shelving system and can store up to 226.8 kg (500 lbs.) per shelf. Additional features include three-point self-latching button closure and reinforced side walls.
Color: Gray
Ordering information: Beige cabinets are also available; contact your Avantor Furniture Specialist for more information.
Spécifications
- Catalog No:
- 15001-720
- 15001-718
- 15001-716
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified