Junkin™ Safety Stretcher Bridle Sling
# de catalogue 56612-685
Fournisseur: Junkin
Small Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Junkin* Safety Stretcher Bridle Sling
- Cat. no.:56612-685
- Supplier No.:JSA-300X
Spécifications
A propos de cet article
For use in rescue situations where stretcher and victim need to be lifted out of danger.
- Provides horizontal or vertical lifting capacity of up to 907 kg (2000 lbs.)
- Made of 2.5 cm (1") nylon webbing with forged nap hooks and Int. Ø. 7.6 cm (3") forged steel suspension ring.
Customers Who Bought This Also Bought