SAFE-T-ZONE® Emergency Stations, Speakman®
Fournisseur: Speakman
CE Compliant
Certificats
A propos de cet article
Emergency stations combine a deluge shower with a eyewash or eye/face wash unit.
- Eyewashes feature dual aerated spray heads with stay-open ball valve
- Eye/face washes feature dual spray heads with automatic flow control
All eyewashes and eye/face washes activate via a stainless steel push handle and include flip-top dust covers. Choose from plastic bowl or stainless steel bowl.
All feature an impeller action deluge showerhead with a high-visibility yellow pull rod, situated on a painted galvanized steel stanchion.
Certifications: Meet ANSI Z358.1 standard.
Customers Who Bought This Also Bought