Lifesaver® Ceiling-Mount Emergency Shower, Speakman®
# de catalogue 14224-248
Fournisseur: Speakman
Small Business Enterprise
Spécifications
- Description:Lifesaver® Ceiling-Mount Emergency Shower
- Cat. no.:14224-248
- Supplier No.:SE-236-PR
Spécifications
A propos de cet article
Lifesaver® emergency shower features a concealed ceiling mounted impeller action chrome plated shower head with a horizontal supply.
Shower is activated with a 2.5 cm (1") stay-open ball valve operated by a high visibility yellow pull rod.
Certifications: Meets ANSI Z358.1 standard.
Customers Who Bought This Also Bought