Emergency Showers, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Designed to mount vertically or horizontally, these emergency showers consist of a 25.4 cm (10") Ø orange ABS plastic showerhead and stainless steel actuating arm, plus a 2.5 cm (1") IPS female inlet.
- Spray pattern is 50.8 cm (20") Ø at 152.4 cm (60") above the floor
- IPS valve is brass with chrome plating or white epoxy/polyester hybrid powder coating
- Pipe nipple on standard construction is orange powder-coated galvanized steel
- Pipe nipple on ColorTech construction (VFCTES647) is brass
Certifications: Meets ANSI Z358.1 standard.
Customers Who Bought This Also Bought