Emergency Shower Tester and Eyewash Test Kit, Speakman®
# de catalogue VF-0950-SE
Fournisseur: Speakman
Spécifications
- Description:Shower and Eyewash test kit
- Cat. no.:VF-0950-SE
- Supplier No.:SE-950
Spécifications
A propos de cet article
Consists of a vinyl-coated rayon sleeve/funnel supported by a handle to direct water flow, plus a 18.9 L (5 gal.) pail to catch water.
- Hoop at end of sleeve fits all standard showers
- Ideal for monthly inspections as required by good laboratory practices
Clear Plexiglas® test kit allows eyewashes to be tested for correct spray pattern.
Customers Who Bought This Also Bought