Lifesaver® Wall-Mount Emergency Showers, Speakman®
Fournisseur: Speakman
Certificats
A propos de cet article
Reach for the handle of these safety showers for immediate rinsing off of harmful contaminants.
- For immediate rinsing off of harmful contaminants
- Impeller action head for more thorough rinsing
- Stay-open valve does not require operator control
An impeller action head allows more thorough rinsing action. The stay-open valve does not require operator control once activated. Showers feature a 20.3 cm (8") Ø yellow shower head, a 2.5 cm (1") NPTF chrome-plated brass ball valve, and a high visibility powder-coated yellow aluminum pull rod. Stainless steel shower head and/or stainless steel pull rod are available as alternative options.
Certifications: Meet ANSI Z358.1 standard.
Customers Who Bought This Also Bought