KEMRESIN® Drop-In Sinks, Kewaunee®
Fournisseur: Kewaunee
Certificats
A propos de cet article
Molded of modified epoxy resin, these durable top-mounted sinks come with coved corners and bottoms pitched to the 3.8 cm (11/2") sized drain outlet.
- Color: black
Sinks meeting ADA requirements are also available.
Customers Who Bought This Also Bought