Oval Cup Sinks, Just Manufacturing
Fournisseur: Just Manufacturing
Certificats
A propos de cet article
For disposal of liquids and distillation
Fabricated of corrosion-resistant 18-gauge, Type 316 stainless steel. Drain consists of an integral crossbar strainer and a 3.8 cm (11/2") IPS stainless steel pipe welded to the sink. Top of sink flange and inside of cup are polished to a hand-blended finish. I.D.: 15.2 L x 7.6 W cm (6 x 3"). Cup sink is self-rimming and includes wing nuts for installation.
Customers Who Bought This Also Bought