Deck-Mounted Gooseneck Single Faucets, with Vacuum Breaker, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Deck-mounted single faucets are made of chrome-plated forged brass and supplied with a serrated hose end, four-arm handle, and 1 cm (3/8") IPS shank, locknut, and washer.
Faucets are available with 15.2 cm (6") or 20.3 cm (8") spread gooseneck with a vacuum breaker. Faucet valve is self-contained renewable unit with replaceable stainless-steel seat.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.
Customers Who Bought This Also Bought