Deck-Mounted Gooseneck Single Faucets, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Deck-mounted single faucets are made of chrome-plated forged brass and supplied with a serrated hose end, four-arm handle and 1 cm (3/8") IPS shank, locknut, and washer.
- Faucet valve is self-contained renewable unit with replaceable stainless steel seat
Faucets are available with 15.2 cm (6") or 20.3 cm (8") spread gooseneck.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.
BodyTextAccessories: Chrome-plated brass faucet accessories include an aerator and an aspirator faucet outlet.
Customers Who Bought This Also Bought