Deck-Mounted Gooseneck Single Faucets, with Nylon Hooded Handles, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Deck-mounted single faucets are made of white epoxy/polyester hybrid powder-coated forged brass.
They are supplied with a serrated hose end, black or color-coded nylon hooded handle, and 1 cm (3/8") IPS shank, locknut, and washer. Faucets are available with 15.2 cm (6") or 20.3 cm (8") spread gooseneck, with or without a vacuum breaker. Faucet valve is self-contained renewable unit with replaceable stainless steel seat.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.
Customers Who Bought This Also Bought