Drench Hoses, WaterSaver Faucet
Fournisseur: WaterSaver Faucet
Certificats
A propos de cet article
Drench hoses provide a soft, wide stream of water that quickly and gently rinses away contaminants.
These feature one FS-Plus Fine Spray head mounted vertically with a self-closing brass squeeze valve. Head has an internal filter, self-regulating flow control, and permanently attached flip-top dust cover.
Certifications: Meet ANSI Z358.1 standard.
Ordering information: Supplied with a 9.5 mm (³/₈") male swivel inlet.
Customers Who Bought This Also Bought