Eyesaver® Deck-Mount Swing-Out Eyewash, Speakman®
# de catalogue 14214-370
Fournisseur: Speakman
Small Business Enterprise
Spécifications
- Description:Eyesaver* Deck-Mount Swing-Out Eyewash
- Cat. no.:14214-370
- Supplier No.:SE-570
Spécifications
A propos de cet article
Low-profile counter-top eyewash unit for installations where space is at a premium.
- Unit is activated when rotated into place
Features twin aerated sprays, stay-open ¹/₄-turn Perfect Valve* with swing activation, polished chrome finish, left or right hand operation for installation flexibility, built-in flow regulator to compensate for varying supply pressures, and deck/wall mounted sign for easy identification.
Certifications: Meets ANSI Z358.1 standard.
Customers Who Bought This Also Bought