SP Bel-Art Transpette™ 8-Channel Transfer Pipet, Disposable, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Perform plate washing, media change, and supernatant collection without the use of pipettors and pipette tips with this inexpensive, 8-channel transfer pipet.
- Disposable
- Available sterile and nonsterile
- Translucent plastic construction
Discard after 'hot' radioactive uses to avoid radiation contamination of expensive liquid handling devices.
Made of translucent plastic, the device holds 600 μl per channel. It cannot be autoclaved. Sterile and non sterile disposable pipetting reservoirs are available.
Ordering information: Non sterile version supplied in a pack of 25. Sterile version individually wrapped in a pack of 100.
Customers Who Bought This Also Bought